Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.52258

Sigma-Aldrich

Fmoc-1-methyl-L-histidine

Novabiochem®

Synonym(s):

Fmoc-1-methyl-L-histidine, Fmoc-1-Methyl-L-Histidine,Fmoc-His(τ-Me)-OH

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$584.00
1 G
$2,250.00

$584.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$584.00
1 G
$2,250.00

About This Item

Empirical Formula (Hill Notation):
C22H21N3O4
CAS Number:
Molecular Weight:
391.42
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

$584.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

product line

Novabiochem®

Assay

≥98.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

[n]1(cnc(c1)C[C@H](NC(=O)OCC2c3c(cccc3)c4c2cccc4)C(=O)O)C

InChI

1S/C22H21N3O4/c1-25-11-14(23-13-25)10-20(21(26)27)24-22(28)29-12-19-17-8-4-2-6-15(17)16-7-3-5-9-18(16)19/h2-9,11,13,19-20H,10,12H2,1H3,(H,24,28)(H,26,27)/t20-/m0/s1

InChI key

SLWOFHRSEVVUHM-FQEVSTJZSA-N

General description

Building block for the incorporation of 1-methylhistidine by Fmoc SPPS.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Application

Fmoc-1-methyl-L-histidine Novabiochem®. CAS 202920-22-7, molar mass 391.42 g/mol.

Analysis Note

Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Assay (HPLC, area%): ≥ 98.0 % (a/a)

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service