Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.51090

Sigma-Aldrich

TFFH

Tetramethylfluoroformamidinium hexafluorophosphate Novabiochem®

Synonym(s):

TFFH, Tetramethylfluoroformamidinium hexafluorophosphate,Fluoro-N,N,N′,N′-bis(tetramethylene)formamidinium hexafluorophosphate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$295.00
25 G
$891.00

$295.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$295.00
25 G
$891.00

About This Item

Empirical Formula (Hill Notation):
C5H12F7N2P
CAS Number:
Molecular Weight:
264.12
MDL number:
UNSPSC Code:
12352107
NACRES:
NA.22

$295.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

product line

Novabiochem®

form

solid

reaction suitability

reaction type: Coupling Reactions

manufacturer/tradename

Novabiochem®

mp

104-109 °C

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

FC(=[N+](C)C)N(C)C

InChI

1S/C5H12FN2/c1-7(2)5(6)8(3)4/h1-4H3/q+1

InChI key

JILUGXGJYPRQPA-UHFFFAOYSA-N

General description

TFFH is an in situ coupling reagents which smoothly converts protected amino acids to the corresponding acid fluoride in the presence of base [1]. Acid fluorides have proven are highly effective at coupling hindered amino acids such as α,α-disubstituted amino acids [2].

Associated Protocols and Technical Articles
Guide to Selection of Coupling Reagents

Literature references

[1] L. A. Carpino, et al. (1995) J. Am. Chem. Soc., 117, 5401.
[2] S. A. Triolo, et al. in ′Peptides 1996, Proc. of 24th European Peptide Symposium′, R. Ramage & R. Epton (Eds), Mayflower Scientific Ltd., Birmingham, 1998, pp. 839.

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Assay 1H-qNMR: ≥ 98.0 %
Ratio Cation/anion in 19F-NMR integration: lot specific result
1:5,7 to 1:6,3
Identity (IR): passes test

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service