Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.51067

Sigma-Aldrich

mono-t-Butoxycarbonyl 1,5-diaminopentane toluenesulfonic acid salt

Novabiochem®

Synonym(s):

mono-t-Butoxycarbonyl 1,5-diaminopentane toluenesulfonic acid salt, Boc-NH(CH 2) 5NH 2 · nTosOH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H22N2O2 · xC7H8O3S
CAS Number:
Molecular Weight:
202.29 (free base basis)
UNSPSC Code:
12352107
NACRES:
NA.22

Quality Level

product line

Novabiochem®

Assay

≥98% (TLC)

form

powder

reaction suitability

reagent type: spacer

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

2-8°C

InChI

1S/C10H22N2O2.C7H8O3S/c1-10(2,3)14-9(13)12-8-6-4-5-7-11;1-6-2-4-7(5-3-6)11(8,9)10/h4-8,11H2,1-3H3,(H,12,13);2-5H,1H3,(H,8,9,10)

InChI key

HIKFZTSCNAAZQN-UHFFFAOYSA-N

General description

Diaminopentane monoprotected with acid-labile Boc protecting group.

Linkage

Replaces: 01-63-0073

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Purity (TLC(0110)): ≥ 98 %
Purity (TLC(CMA2)): ≥ 98 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service