Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.51025

Sigma-Aldrich

5-Carboxyfluorescein

Novabiochem®

Synonym(s):

5-Carboxyfluorescein, 5-FAM

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$56.20
100 MG
$168.00
1 G
$1,250.00
5 G
$4,350.00

$56.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$56.20
100 MG
$168.00
1 G
$1,250.00
5 G
$4,350.00

About This Item

Empirical Formula (Hill Notation):
C21H12O7
CAS Number:
Molecular Weight:
376.32
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.22

$56.20


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

product line

Novabiochem®

Assay

≥98% (TLC)
≥99.0% (HPLC)

form

powder

reaction suitability

reagent type: fluorescent-labelling reagent

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

O1C3(c4c(cc(cc4)O)Oc5c3ccc(c5)O)c2c(cc(cc2)C(=O)O)C1=O

InChI

1S/C21H12O7/c22-11-2-5-15-17(8-11)27-18-9-12(23)3-6-16(18)21(15)14-4-1-10(19(24)25)7-13(14)20(26)28-21/h1-9,22-23H,(H,24,25)

InChI key

NJYVEMPWNAYQQN-UHFFFAOYSA-N

General description

This high purity fluorescent dye (λex494 nm, λem 518 nm) is supplied as a single isomer, which ensures production of labeled products of defined chemical structure, as well as greatly simplifying purification and characterization. FAM should be introduced as the last step in the synthesis to avoid formation of side-products, or be protected by tritylation prior to acylation or hydrazine treatment [1].

Associated Protocols and Technical Articles
Chromogenic Labeling of Synthetic Peptides

Literature references

[1] R. Fischer, et al. (2003) Bioconjugate Chem., 14, 653.

Application

5-Carboxyfluorescein Novabiochem®. CAS 76823-03-5, molar mass 376.32 g/mol.

Linkage

Replaces: 01-63-0112

Analysis Note

Colour (visual): yellow to orange
Appearance of substance (visual): powder
Identity (IR): passes test
Purity (TLC (EtOAc/MeOH/H2O/DIPEA 60 : 30 : 10 : 3)): ≥ 98 %
Assay (HPLC, area%): ≥ 99.0 % (a/a)

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Chromogenic and fluorogenic derivatives are invaluable tools for biochemistry, having numerous applications in enzymology, protein chemistry, immunology and histochemistry.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service