Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.51003

Sigma-Aldrich

4-(4-Formyl-3,5-dimethoxyphenoxy)butyric acid

Novabiochem®

Synonym(s):

4-(4-Formyl-3,5-dimethoxyphenoxy)butyric acid, BAL-linker

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16O6
CAS Number:
Molecular Weight:
268.26
UNSPSC Code:
12352106
Pricing and availability is not currently available.

Quality Level

product line

Novabiochem®

Assay

≥97.0% (HPLC)
≥98% (TLC)

form

powder

reaction suitability

reagent type: linker

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

2-8°C

InChI

1S/C13H16O6/c1-17-11-6-9(19-5-3-4-13(15)16)7-12(18-2)10(11)8-14/h6-8H,3-5H2,1-2H3,(H,15,16)

InChI key

SEJSVFHBVLKHLB-UHFFFAOYSA-N

General description

BAL-linker. Very acid sensitive linker for the preparation of carboxamides, carbamates and C-terminally modified peptides [1-4]

The product number for this product was previously 01-60-0086.

To obtain a certificate of analysis (CoA) of a lot that begins with the letter “A”, please select the option in the right hand menu “Request a COA for Lot#s starting with A”.

Linkage

Replaces: 01-60-0086

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Purity (TLC(157A)): ≥ 98 %
Assay (HPLC, area%): ≥ 97.0 % (a/a)

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service