Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.45152

Sigma-Aldrich

Dimethyl carbonate

Msynthplus

Synonym(s):

Dimethyl carbonate, DMC, Methyl carbonate, Carbonic acid dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2CO
CAS Number:
Molecular Weight:
90.08
MDL number:
UNSPSC Code:
12352108
EC Index Number:
210-478-4

vapor pressure

53 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

458 °C

potency

9000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

expl. lim.

4.22-12.87 % (v/v)

bp

90 °C/1016 hPa

mp

0.5-4.7 °C

transition temp

flash point 16.7 °C

density

1.07 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C3H6O3/c1-5-3(4)6-2/h1-2H3

InChI key

IEJIGPNLZYLLBP-UHFFFAOYSA-N

Analysis Note

Appearance of substance (visual): liquid
Colour number (Hazen): ≤ 20
Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.068 - 1.070
Identity (IR): passes test

Legal Information

MSYNTH is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(C)

16 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service