Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.45097

Sigma-Aldrich

N-Dodecane

Msynthplus

Synonym(s):

N-Dodecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10CH3
CAS Number:
Molecular Weight:
170.33
MDL number:
UNSPSC Code:
12352001
EC Index Number:
203-967-9

vapor pressure

0.4 hPa ( 20 °C)

Quality Level

Assay

≥99% (GC)

form

liquid

autoignition temp.

200 °C

expl. lim.

0.6 % (v/v)

kinematic viscosity

<7 cSt(40 °C)

bp

216.3 °C/1013 hPa

mp

-9.6 °C

transition temp

flash point 85.5 °C

density

0.75 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C12H26/c1-3-5-7-9-11-12-10-8-6-4-2/h3-12H2,1-2H3

InChI key

SNRUBQQJIBEYMU-UHFFFAOYSA-N

Analysis Note

Color (visual): colourless
Appearance of substance (visual): liquid
Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.748 - 0.749
Identity (IR): passes test
1,1-Dichloroethene (HS-GC): lot specific result
1,1,1-Trichloroethane (HS-GC): lot specific result
Benzene (HS-GC): lot specific result
Carbon tetrachloride (HS-GC): lot specific result
1,2-Dichloroethane (HS-GC): lot specific result

Legal Information

MSYNTH is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(C)

70 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service