Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.45061

Sigma-Aldrich

Triethylamine

Synonym(s):

Triethylamine, N,N-Diethylethanamine, TEA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3N
CAS Number:
Molecular Weight:
101.19
MDL number:
UNSPSC Code:
12352116

vapor pressure

72 hPa ( 20 °C)

Quality Level

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

215 °C

potency

730 mg/kg LD50, oral (Rat)

expl. lim.

1.2-9.3 % (v/v)

pH

12.7 (15 °C, 100 g/L in H2O, IUCLID)

bp

90 °C/1013 hPa

mp

-115 °C

transition temp

flash point -11 °C

density

0.73 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C6H15N/c1-4-7(5-2)6-3/h4-6H2,1-3H3

InChI key

ZMANZCXQSJIPKH-UHFFFAOYSA-N

Analysis Note

Appearance of substance (visual): clear
Colour number: ≤ 20
Assay (GC, area%): ≥ 99.5 % (a/a)
Density (d 20 °C/ 4 °C): 0.726 - 0.728
Water (K. F.): ≤ 0.20 %
Identity (IR): passes test

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service