Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.45008

Sigma-Aldrich

Selenium dioxide

(sublimed) Msynthplus

Synonym(s):

Selenium dioxide, Selenious anhydride, Selenium(IV) oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
O2Se
CAS Number:
Molecular Weight:
110.96
UNSPSC Code:
12352303
EC Index Number:
231-194-7

vapor pressure

<1 Pa ( 20 °C)

form

powder

potency

68.1 mg/kg LD50, oral (Rat)

reaction suitability

reagent type: oxidant

pH

2 (20 °C, 10 g/L in H2O)

mp

340 °C

solubility

1353 g/L

density

3.95 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/O2Se/c1-3-2

InChI key

JPJALAQPGMAKDF-UHFFFAOYSA-N

Analysis Note

Colour (visual): white
Appearance of substance (visual): powder
Assay (iodometric): ≥ 98.0 %
Te (Tellur) (ICP MS): lot specific result

Legal Information

MSYNTH is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service