Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.44008

Sigma-Aldrich

tert-Butyl methyl ether

for synthesis

Synonym(s):

tert-Butyl methyl ether, MTBE, MTB, 2-Methoxy-2-methylpropane, Methyl tert-butyl ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
88.15
MDL number:
UNSPSC Code:
12352112
EC Index Number:
216-653-1
grade:
for synthesis
synthesis grade
bp:
55.3 °C/1013 hPa
vapor pressure:
268 hPa ( 20 °C)

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

268 hPa ( 20 °C)

form

liquid

autoignition temp.

460 °C

potency

>2000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.5-8.5 % (v/v)

dilution

(for synthesis)

kinematic viscosity

0.409 cSt(40 °C)

bp

55.3 °C/1013 hPa

mp

-108.6 °C

transition temp

flash point -28 °C

solubility

42 g/L

density

0.74 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C5H12O/c1-5(2,3)6-4/h1-4H3

InChI key

BZLVMXJERCGZMT-UHFFFAOYSA-N

General description

for synthesis

Application

tert-Butyl methyl ether for synthesis. CAS 1634-04-4, EC Number 216-653-1, chemical formula (CH₃)₃COCH₃.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-18.4 °F - closed cup

Flash Point(C)

-28 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service