Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.43983

Sigma-Aldrich

Benzethonium chloride

for synthesis

Synonym(s):

Benzethonium chloride, N-Benzyl-N,N-dimethyl-N-[4-(1.1.3.3-tetramethylbutyl)-phenoxyethoxyethyl]ammonium chloride, Benzethonium chloride

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$395.00

$395.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 G
$395.00

About This Item

Empirical Formula (Hill Notation):
C27H42ClNO2
CAS Number:
Molecular Weight:
448.08
MDL number:
UNSPSC Code:
12352101
EC Index Number:
204-479-9
NACRES:
NA.22

$395.00


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

<0.001 hPa ( 25 °C)

Quality Level

form

solid

autoignition temp.

380 °C

potency

295 mg/kg LD50, oral (Rat)

pH

5-6.5 (10 g/L in H2O)

mp

160-166 °C

bulk density

440 kg/m3

storage temp.

15-25°C

SMILES string

[Cl-].[N+](CCOCCOc2ccc(cc2)C(CC(C)(C)C)(C)C)(Cc1ccccc1)(C)C

InChI

1S/C27H42NO2.ClH/c1-26(2,3)22-27(4,5)24-13-15-25(16-14-24)30-20-19-29-18-17-28(6,7)21-23-11-9-8-10-12-23;/h8-16H,17-22H2,1-7H3;1H/q+1;/p-1

InChI key

UREZNYTWGJKWBI-UHFFFAOYSA-M

Application

  • Development of biologically active ionic liquids: Research highlighted the synthesis of new ionic liquids incorporating the benzethonium cation, which show promising SAR induction and antimicrobial properties, potentially useful in pest management and disease control applications (Czerwoniec et al., 2024).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service