Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.43942

Sigma-Aldrich

Propanephosphonic acid anhydride

(50% solution in ethyl acetate) for synthesis T3P

Synonym(s):

2,4,6-Tripropyl-1,3,5,2,4,6-trioxatriphosphorinane-2,4,6-trioxide solution, PPACA, Propylphosphonic anhydride, T3P

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$101.00
50 ML
$161.00
100 ML
$263.00
2.5 L
$6,810.00

$101.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
$101.00
50 ML
$161.00
100 ML
$263.00
2.5 L
$6,810.00

About This Item

UNSPSC Code:
12352108
NACRES:
NA.22

$101.00


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

97 hPa ( 20 °C)

Quality Level

form

liquid

bp

77 °C/1013 hPa (ethyl acetate)

transition temp

flash point -4 °C (ethyl acetate)

density

1.08 g/cm3 at 20 °C

storage temp.

2-8°C

Application

  • Cyclization of Linear Tetrapeptides Containing N‐Methylated Amino Acids: The study reports the synthesis of naturally occurring cyclotetrapeptides using 1-propanephosphonic acid anhydride, noting unexpected findings in cyclization patterns. (Zhang et al., 2017).

Analysis Note

Assay (T3P) (NMR): ≥ 40.0 %
Pyropropanephosphonic acid (NMR): ≤ 10.0 %
Propanephosphonic acid (NMR): ≤ 0.50 %
Ethyl acetate (calculated): ≤ 50.0 %

Signal Word

Danger

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

24.8 °F - ethyl acetate

Flash Point(C)

-4 °C - ethyl acetate


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service