Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.43851

Sigma-Aldrich

3-Benzoylpyridine

≥98% (GC)

Synonym(s):

3-Benzoylpyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9NO
CAS Number:
Molecular Weight:
183.21
UNSPSC Code:
12352115
EC Index Number:
226-561-3
NACRES:
NA.22

Quality Level

Assay

≥98% (GC)

form

solid

storage temp.

2-30°C

InChI

1S/C12H9NO/c14-12(10-5-2-1-3-6-10)11-7-4-8-13-9-11/h1-9H

InChI key

RYMBAPVTUHZCNF-UHFFFAOYSA-N

General description

for synthesis

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 36 °C
Melting range (upper value): ≤ 40 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service