Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.43805

Sigma-Aldrich

Triisooctylamine

for synthesis

Synonym(s):

Triisooctylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C8H17)3N
CAS Number:
Molecular Weight:
353.67
MDL number:
UNSPSC Code:
12352116
EC Index Number:
247-092-0

Assay

≥95.0% (acidimetric)

form

liquid

bp

>204 °C

transition temp

flash point 110 °C (DIN 51758)

density

0.800 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C24H51N/c1-22(2)16-10-7-13-19-25(20-14-8-11-17-23(3)4)21-15-9-12-18-24(5)6/h22-24H,7-21H2,1-6H3

InChI key

YKGBNAGNNUEZQC-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 95.0 %
Density (d 20 °C/ 4 °C): 0.814 - 0.818
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service