Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.43410

Sigma-Aldrich

Triethyl phosphonoformate

≥97%

Synonym(s):

Triethyl phosphonoformate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15O5P
CAS Number:
Molecular Weight:
210.16
MDL number:
UNSPSC Code:
12352108
EC Index Number:
216-016-8

Assay

≥97%

storage temp.

2-30°C

InChI

1S/C7H15O5P/c1-4-10-7(8)13(9,11-5-2)12-6-3/h4-6H2,1-3H3

InChI key

NOJFJZZMRDSOLM-UHFFFAOYSA-N

General description

for synthesis

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.121 - 1.125
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service