Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.42258

Sigma-Aldrich

2-(4-Toluoyl)benzoic acid

for synthesis

Synonym(s):

2-(4-Toluoyl)benzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$84.50

$84.50


Please contact Customer Service for Availability


Select a Size

Change View
25 G
$84.50

About This Item

Empirical Formula (Hill Notation):
C15H12O3
CAS Number:
Molecular Weight:
240.25
MDL number:
UNSPSC Code:
12352106
EC Index Number:
201-614-3
NACRES:
NA.22

$84.50


Please contact Customer Service for Availability

Quality Level

Assay

≥97.0% (acidimetric)

mp

133-137 °C

storage temp.

2-30°C

SMILES string

OC(=O)c1c(cccc1)C(=O)c2ccc(cc2)C

InChI

1S/C15H12O3/c1-10-6-8-11(9-7-10)14(16)12-4-2-3-5-13(12)15(17)18/h2-9H,1H3,(H,17,18)

InChI key

ICQOWIXIHDDXDI-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 97.0 %
Melting range (lower value): ≥ 133 °C
Melting range (upper value): ≤ 137 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service