Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.42141

Sigma-Aldrich

1,5-Dichloropentane

for synthesis

Synonym(s):

1,5-Dichloropentane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10Cl2
CAS Number:
Molecular Weight:
141.04
MDL number:
UNSPSC Code:
12352101
EC Index Number:
211-053-6
NACRES:
NA.22

vapor pressure

1.5 hPa ( 25 °C)

Quality Level

Assay

≥98.0% (GC)

bp

180 °C/1013 hPa

mp

-72 °C

transition temp

flash point 26 °C

solubility

0.125 g/L

density

1.10 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C5H10Cl2/c6-4-2-1-3-5-7/h1-5H2

InChI key

LBKDGROORAKTLC-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.098 - 1.103
Identity (IR): passes test

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service