Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.42025

Sigma-Aldrich

Trichloro(chloromethyl)silane

for synthesis

Synonym(s):

Trichloro(chloromethyl)silane

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$159.00

$159.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
$159.00

About This Item

CAS Number:
Molecular Weight:
183.93
MDL number:
UNSPSC Code:
12352103
EC Index Number:
216-316-9
NACRES:
NA.22

$159.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

liquid

bp

117-118 °C/1013 hPa

transition temp

flash point 69 °C

density

1.47 g/cm3

storage temp.

2-30°C

SMILES string

[Si](Cl)(Cl)(Cl)CCl

InChI

1S/CH2Cl4Si/c2-1-6(3,4)5/h1H2

InChI key

FYTPGBJPTDQJCG-UHFFFAOYSA-N

General description

for synthesis

Application

Trichloro(chloromethyl)silane for synthesis. CAS 1558-25-4, molar mass 183.93 g/mol.

Analysis Note

Assay (GC, area%)≥ 95.5 %Identity (IR)passes test

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

156.2 °F

Flash Point(C)

69 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service