Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.41865

Sigma-Aldrich

Thiophene-2-acetyl chloride

for synthesis

Synonym(s):

Thiophene-2-acetyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
160.62
MDL number:
UNSPSC Code:
12352106
EC Index Number:
254-290-0

Quality Level

form

liquid

bp

130-135 °C/120 hPa

transition temp

flash point 101 °C

density

1.303 g/cm3 at 20 °C

storage temp.

2-8°C

InChI

1S/C6H5ClOS/c7-6(8)4-5-2-1-3-9-5/h1-3H,4H2

InChI key

AJYXPNIENRLELY-UHFFFAOYSA-N

General description

for synthesis

Application

Thiophene-2-acetyl chloride for synthesis. CAS 39098-97-0, molar mass 160.62 g/mol.

Analysis Note

Assay (GC, area%)≥ 96.0 % (a/a)Density (d 20 °C/ 4 °C)1.300 - 1.305Identity (IR)passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(C)

102 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service