Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.41858

Sigma-Aldrich

1,4-Dioxa-8-azaspiro[4.5]decane

for synthesis

Synonym(s):

1,4-Dioxa-8-azaspiro[4.5]decane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13NO2
CAS Number:
Molecular Weight:
143.18
MDL number:
UNSPSC Code:
12352005
EC Index Number:
205-868-6
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (GC)

form

liquid

bp

108-110 °C/35 hPa

transition temp

flash point 81 °C

density

1.11 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C7H13NO2/c1-3-8-4-2-7(1)9-5-6-10-7/h8H,1-6H2/p+1

InChI key

KPKNTUUIEVXMOH-UHFFFAOYSA-O

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.110 - 1.115
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(C)

81 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service