Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.41772

Sigma-Aldrich

3-Amino-4-chlorobenzotrifluoride

for synthesis

Synonym(s):

3-Amino-4-chlorobenzotrifluoride

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$149.00

$149.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
$149.00

About This Item

Empirical Formula (Hill Notation):
C7H5ClF3N
CAS Number:
Molecular Weight:
195.57
MDL number:
UNSPSC Code:
12352116
EC Index Number:
204-475-7
NACRES:
NA.22

$149.00


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

0.15 hPa

Quality Level

Assay

≥97.0% (GC)

form

liquid

autoignition temp.

≥560 °C

bp

82-83 °C/12 hPa

mp

10 °C

transition temp

flash point 75 °C

solubility

11 g/L

density

1.428 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

FC(F)(F)c1cc(c(cc1)Cl)N

InChI

1S/C7H5ClF3N/c8-5-2-1-4(3-6(5)12)7(9,10)11/h1-3H,12H2

InChI key

VKTTYIXIDXWHKW-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.426 - 1.430
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service