Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.41501

Sigma-Aldrich

2-Sulfobenzoic anhydride

for synthesis

Synonym(s):

2-Sulfobenzoic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4O4S
CAS Number:
Molecular Weight:
184.17
MDL number:
UNSPSC Code:
12352115
EC Index Number:
201-322-6
NACRES:
NA.22

Quality Level

mp

116-122 °C

storage temp.

2-30°C

InChI

1S/C7H4O4S/c8-7-5-3-1-2-4-6(5)12(9,10)11-7/h1-4H

InChI key

NCYNKWQXFADUOZ-UHFFFAOYSA-N

Analysis Note

Assay (morpholine method): ≥ 96.0 %
Melting range (lower value): ≥ 117 °C
Melting range (upper value): ≤ 127 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service