Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.41435

Sigma-Aldrich

4-(4-Nitrophenylazo)-1-naphthol

for synthesis

Synonym(s):

4-(4-Nitrophenylazo)-1-naphthol, Magneson II

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$228.00

$228.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$228.00

About This Item

Empirical Formula (Hill Notation):
C16H11N3O3
CAS Number:
Molecular Weight:
293.28
MDL number:
UNSPSC Code:
12352104
EC Index Number:
226-129-4
NACRES:
NA.22

$228.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

solid

mp

277-280 °C (decomposition)

storage temp.

2-30°C

SMILES string

[N+](=O)([O-])c1ccc(cc1)N\N=C2\c3c(cccc3)C(=O)C=C\2

InChI

1S/C16H11N3O3/c20-16-10-9-15(13-3-1-2-4-14(13)16)18-17-11-5-7-12(8-6-11)19(21)22/h1-10,17H/b18-15+

InChI key

UYDXOFSMBKSLPM-OBGWFSINSA-N

Analysis Note

Assay (TLC): ≥ 90.0 %
Melting range (lower value): ≥ 278 °C
Melting range (upper value): ≤ 284 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service