Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.41321

Sigma-Aldrich

Citraconic anhydride

for synthesis

Synonym(s):

Citraconic anhydride, Methylmaleic anhydride

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$113.00

$113.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$113.00

About This Item

Empirical Formula (Hill Notation):
C5H4O3
CAS Number:
Molecular Weight:
112.08
MDL number:
UNSPSC Code:
12352115
EC Index Number:
210-459-0
NACRES:
NA.22

$113.00


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

1.3 hPa ( 47 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

2600 mg/kg LD50, oral (Rat)
218 mg/kg LD50, skin (Rabbit)

bp

213-214 °C/1013 hPa

mp

7-8 °C

transition temp

flash point 111 °C

density

1.243 g/cm3 at 20 °C

SMILES string

O1C(=O)C(=CC1=O)C

InChI

1S/C5H4O3/c1-3-2-4(6)8-5(3)7/h2H,1H3

InChI key

AYKYXWQEBUNJCN-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.241 - 1.246
Identity (IR): passes test

Pictograms

CorrosionHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

231.8 °F

Flash Point(C)

111 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service