Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.41278

Sigma-Aldrich

5,6-Dimethylbenzimidazole

for synthesis

Synonym(s):

5,6-Dimethylbenzimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10N2
CAS Number:
Molecular Weight:
146.19
MDL number:
UNSPSC Code:
12352005
EC Index Number:
209-488-1

Assay

≥99.0% (HPLC)

mp

202-206 °C

storage temp.

2-30°C

InChI

1S/C9H10N2/c1-6-3-8-9(4-7(6)2)11-5-10-8/h3-5H,1-2H3,(H,10,11)

InChI key

LJUQGASMPRMWIW-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 203 °C
Melting range (upper value): ≤ 206 °C
Identity (IR): passes test

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service