Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.41188

Sigma-Aldrich

Tetra-n-butylammonium borohydride

for synthesis

Synonym(s):

Tetra-n-butylammonium borohydride, Tetra-n-butylammonium-tetrahydridoborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H40BN
CAS Number:
Molecular Weight:
257.31
MDL number:
UNSPSC Code:
12352107
EC Index Number:
251-658-2
Pricing and availability is not currently available.

form

crystals

reaction suitability

reagent type: reductant

mp

124-129 °C

transition temp

flash point 140 °C

storage temp.

2-30°C

InChI

1S/C16H36N.BH4/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;/h5-16H2,1-4H3;1H4/q+1;-1

InChI key

GMBOFJFPOCGSOI-UHFFFAOYSA-N

Analysis Note

Assay (hypochlorite): ≥ 97.0 %
Identity (IR): passes test

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Water-react. 2

Storage Class Code

4.3 - Hazardous materials, which set free flammable gases upon contact with water

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service