Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.40126

Sigma-Aldrich

Butyltinhydroxide-oxide

for synthesis

Synonym(s):

Butyltinhydroxide-oxide, Butylhydroxyoxo stannane

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$66.50
100 G
$655.00

$66.50


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$66.50
100 G
$655.00

About This Item

Empirical Formula (Hill Notation):
C4H10O2Sn
CAS Number:
Molecular Weight:
208.83
MDL number:
UNSPSC Code:
12352103
EC Index Number:
218-880-1
NACRES:
NA.22

$66.50


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

1.47 hPa ( 20 °C)

Quality Level

form

powder

bp

>350 °C/1013 hPa

storage temp.

2-30°C

SMILES string

[Sn](=O)(O)CCCC

InChI

1S/C4H9.H2O.O.Sn/c1-3-4-2;;;/h1,3-4H2,2H3;1H2;;/q;;;+1/p-1

InChI key

WIHMDCQAEONXND-UHFFFAOYSA-M

Analysis Note

Assay (Sn): 54.0 - 60.0 %
Identity (IR): passes test

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service