Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.24508

Sigma-Aldrich

(+)-trans-2-tert-Butylcyclohexanol

for synthesis

Synonym(s):

(+)-trans-2-tert-Butylcyclohexanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O
CAS Number:
Molecular Weight:
156.27
UNSPSC Code:
12352104

Assay

≥99.0% (GC)

form

crystals

bp

88 °C/10.7 hPa

mp

54-57 °C

storage temp.

2-30°C

InChI

1S/C10H20O/c1-10(2,3)8-6-4-5-7-9(8)11/h8-9,11H,4-7H2,1-3H3/t8-,9-/m1/s1

InChI key

DLTWBMHADAJAAZ-RKDXNWHRSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 54 °C
Melting range (upper value): ≤ 57 °C
Spec. rotation [α²0/D (c=1 in chloroform): +40.0 - +44.0 °
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service