Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.22145

Sigma-Aldrich

Sodium sulfanilate dihydrate

for synthesis

Synonym(s):

Sodium sulfanilate dihydrate, Sulfanilic acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$126.00

$126.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 G
$126.00

About This Item

Linear Formula:
4-(NH2)C6H4SO3Na · 2H2O
CAS Number:
Molecular Weight:
231.20
MDL number:
UNSPSC Code:
12352107
EC Index Number:
208-208-5
NACRES:
NA.22

$126.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

powder

solubility

170 g/L

storage temp.

2-30°C

SMILES string

[Na+].[S](=O)(=O)([O-])c1ccc(cc1)N.O.O

InChI

1S/C6H7NO3S.Na.2H2O/c7-5-1-3-6(4-2-5)11(8,9)10;;;/h1-4H,7H2,(H,8,9,10);;2*1H2/q;+1;;/p-1

InChI key

MJFZIDUBUNIFLZ-UHFFFAOYSA-M

Analysis Note

Assay (ex Na): ≥ 98.0 %
Water (K. F.): 15.0 - 17.0 %
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service