Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.22063

Sigma-Aldrich

1-Hexadecanethiol

for synthesis

Synonym(s):

1-Hexadecanethiol, Cetyl mercaptan, Hexadecyl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15SH
CAS Number:
Molecular Weight:
258.51
MDL number:
UNSPSC Code:
12352105
EC Index Number:
220-846-6
NACRES:
NA.22

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

Assay

≥88.0% (GC)

form

liquid

autoignition temp.

260 °C

mp

18-20 °C

transition temp

flash point 102 °C

density

0.85 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C16H34S/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h17H,2-16H2,1H3

InChI key

ORTRWBYBJVGVQC-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 88.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.845 - 0.850
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

215.6 °F

Flash Point(C)

102 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service