Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.22058

Sigma-Aldrich

Gluconic acid sodium salt

for synthesis

Synonym(s):

Gluconic acid sodium salt, Sodium gluconate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$39.10
1 KG
$64.50
2.5 KG
$124.00
25 KG
$540.00
50 KG
$1,530.00

$39.10


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$39.10
1 KG
$64.50
2.5 KG
$124.00
25 KG
$540.00
50 KG
$1,530.00

About This Item

Empirical Formula (Hill Notation):
C6H11NaO7
CAS Number:
Molecular Weight:
218.14
MDL number:
UNSPSC Code:
12352107
EC Index Number:
208-407-7
NACRES:
NA.22

$39.10


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

powder

autoignition temp.

>200 °C

potency

>2000 mg/kg LD50, oral (Rat)

pH

7.0-8.0 (20 °C, 100 g/L in H2O)

mp

170-175 °C

solubility

590 g/L

storage temp.

2-30°C

SMILES string

[Na+].[O-]C(=O)C(O)C(O)C(O)C(O)CO

InChI

1S/C6H12O7.Na/c7-1-2(8)3(9)4(10)5(11)6(12)13;/h2-5,7-11H,1H2,(H,12,13);/q;+1/p-1

InChI key

UPMFZISCCZSDND-UHFFFAOYSA-M

Related Categories

Analysis Note

Assay (HClO₄): ≥ 99.0 %
Spec. rotation [α²0/D (c=10 in water): +11.5 - +12.5 °
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service