Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.22048

Sigma-Aldrich

1-Dodecene

for synthesis

Synonym(s):

1-Dodecene, 1-Dodecylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9CH=CH2
CAS Number:
Molecular Weight:
168.32
MDL number:
UNSPSC Code:
12352002
EC Index Number:
203-968-4
NACRES:
NA.22

vapor pressure

0.193 hPa ( 25 °C)

Quality Level

Assay

≥94.0% (GC)

form

liquid

autoignition temp.

226 °C

potency

>10000 mg/kg LD50, oral (Rat)
>10000 mg/kg LD50, skin (Rabbit)

expl. lim.

0.6-5.4 % (v/v)

kinematic viscosity

0.68 cSt(100 °C)

bp

213-216 °C/1013 hPa

mp

-35 °C

transition temp

flash point 77 °C

density

0.76 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C12H24/c1-3-5-7-9-11-12-10-8-6-4-2/h3H,1,4-12H2,2H3

InChI key

CRSBERNSMYQZNG-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 94.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.758 - 0.759
Identity (IR): passes test

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

170.6 °F

Flash Point(C)

77 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service