Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.22046

Sigma-Aldrich

Docosane

for synthesis

Synonym(s):

Docosane

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$108.00

$108.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 G
$108.00

About This Item

Linear Formula:
CH3(CH2)20CH3
CAS Number:
Molecular Weight:
310.60
MDL number:
UNSPSC Code:
12352001
EC Index Number:
211-121-5
NACRES:
NA.22

$108.00


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

<1 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

solid

bp

369 °C/1013 hPa

mp

44 °C

transition temp

flash point >110 °C

density

0.794 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

C(CCCCCCCCCCC)CCCCCCCCCC

InChI

1S/C22H46/c1-3-5-7-9-11-13-15-17-19-21-22-20-18-16-14-12-10-8-6-4-2/h3-22H2,1-2H3

InChI key

HOWGUJZVBDQJKV-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 42 °C
Melting range (upper value): ≤ 45 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service