Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.22040

Sigma-Aldrich

N,N-Dimethyl-p-toluidine

for synthesis

Synonym(s):

N,N-Dimethyl-p-toluidine, 4-Dimethylaminotoluene

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$36.60
1 L
$197.00
25 KG
$1,410.00

$36.60


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
$36.60
1 L
$197.00
25 KG
$1,410.00

About This Item

Linear Formula:
4-(CH3)C6H4N(CH3)2
CAS Number:
Molecular Weight:
135.21
MDL number:
UNSPSC Code:
12352116
EC Index Number:
202-805-4
NACRES:
NA.22

$36.60


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

0.1 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

oil

autoignition temp.

425 °C

potency

1650 mg/kg LD50, oral (Rat)

bp

215 °C/1013 hPa

mp

-15 °C

transition temp

flash point 76 °C

density

0.94 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(C)(C)c1ccc(cc1)C

InChI

1S/C9H13N/c1-8-4-6-9(7-5-8)10(2)3/h4-7H,1-3H3

InChI key

GYVGXEWAOAAJEU-UHFFFAOYSA-N

Application



  • Zinc catalysed electrophilic C–H borylation of heteroarenes: This article explores a zinc-catalyzed process for borylation of heteroarenes using N,N-dimethyl-p-toluidine (Grundy et al., 2021).


  • C–H bond sulfonylation of anilines with the insertion of sulfur dioxide under metal-free conditions: Details a sulfonylation reaction involving N,N-dimethyl-p-toluidine under metal-free conditions (Zhou et al., 2018).



Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.935 - 0.937
Identity (IR): passes test

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 3 - STOT RE 2 Oral

Target Organs

Reproductive organs

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service