Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.22010

Sigma-Aldrich

1-Decen

for synthesis

Synonym(s):

1-Decen, 1-Decylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH2
CAS Number:
Molecular Weight:
140.27
MDL number:
UNSPSC Code:
12352002
EC Index Number:
212-819-2

vapor pressure

1.4 hPa ( 20 °C)

Quality Level

autoignition temp.

210 °C

potency

>10000 mg/kg LD50, oral (Rat)
>10000 mg/kg LD50, skin (Rabbit)

expl. lim.

0.7-5.9 % (v/v)

refractive index

n20/589 1.4214

kinematic viscosity

1.1 cSt(20 °C)

bp

170.5 °C/1013 hPa

mp

-66 °C

transition temp

flash point 49 °C

density

0.74 g/cm3 at 20 °C

InChI

1S/C10H20/c1-3-5-7-9-10-8-6-4-2/h3H,1,4-10H2,2H3

InChI key

AFFLGGQVNFXPEV-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service