Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.21958

Sigma-Aldrich

Propyl benzoate

for synthesis

Synonym(s):

Propyl benzoate, Benzoic acid propyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$36.70
250 ML
$104.00

$36.70


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
$36.70
250 ML
$104.00

About This Item

Empirical Formula (Hill Notation):
C10H12O2
CAS Number:
Molecular Weight:
164.20
MDL number:
UNSPSC Code:
12352108
EC Index Number:
219-020-8
NACRES:
NA.22

$36.70


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

0.181 hPa ( 25 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

bp

231 °C/1013 hPa

mp

-52 °C

transition temp

flash point >100 °C

density

1.02 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CCC)C(=O)c1ccccc1

InChI

1S/C10H12O2/c1-2-8-12-10(11)9-6-4-3-5-7-9/h3-7H,2,8H2,1H3

InChI key

UDEWPOVQBGFNGE-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.023 - 1.025
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

212.0 °F

Flash Point(C)

> 100 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service