Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.21770

Sigma-Aldrich

1,2:7,8-Diepoxyoctane

for synthesis

Synonym(s):

1,2:7,8-Diepoxyoctane, 1,7-Octadiene dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O2
CAS Number:
Molecular Weight:
142.20
MDL number:
UNSPSC Code:
12352005
EC Index Number:
219-375-9

vapor pressure

<1 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

350 °C (DIN 51794)

potency

1049 mg/kg LD50, oral (Rat)
314 mg/kg LD50, skin (Rabbit)

bp

222 °C/1013 hPa

mp

7 °C

transition temp

flash point 89 °C (DIN 51758)

solubility

7 g/L

density

0.98 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C8H14O2/c1(3-7-5-9-7)2-4-8-6-10-8/h7-8H,1-6H2

InChI key

LFKLPJRVSHJZPL-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.983 - 0.985
Identity (IR): passes test

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Carc. 1B - Muta. 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service