Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.21667

Sigma-Aldrich

Trichloroethylsilane

for synthesis

Synonym(s):

Trichloroethylsilane, Ethyltrichlorosilane

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$215.00

$215.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
$215.00

About This Item

Empirical Formula (Hill Notation):
C2H5Cl3Si
CAS Number:
Molecular Weight:
163.51
MDL number:
UNSPSC Code:
12352103
EC Index Number:
204-072-6
NACRES:
NA.22

$215.00


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

36 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

385 °C

potency

1330 mg/kg LD50, oral (Rat)

expl. lim.

2.68 % (v/v)

bp

98 °C/1013 hPa

mp

-105 °C

transition temp

flash point 14 °C

density

1.24 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Si](Cl)(Cl)(Cl)CC

InChI

1S/C2H5Cl3Si/c1-2-6(3,4)5/h2H2,1H3

InChI key

ZOYFEXPFPVDYIS-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 2 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service