Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.21196

Sigma-Aldrich

Triphenylmethanol

for synthesis

Synonym(s):

Triphenylmethanol, Triphenyl carbinol

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$99.00
250 G
$249.00

$99.00


Available to ship onApril 03, 2025Details


Request a Bulk Order

Select a Size

Change View
50 G
$99.00
250 G
$249.00

About This Item

Linear Formula:
(C6H5)3COH
CAS Number:
Molecular Weight:
260.33
MDL number:
UNSPSC Code:
12352104
EC Index Number:
200-988-5
NACRES:
NA.22

$99.00


Available to ship onApril 03, 2025Details


Request a Bulk Order

Quality Level

Assay

≥98.0% (HPLC)

form

powder

mp

160-163 °C (External MSDS)

solubility

insoluble 1.43 g/L

storage temp.

2-30°C

SMILES string

OC(c3ccccc3)(c2ccccc2)c1ccccc1

InChI

1S/C19H16O/c20-19(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15,20H

InChI key

LZTRCELOJRDYMQ-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service