Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.21180

Sigma-Aldrich

Trimethyl borate

for synthesis

Synonym(s):

Trimethyl borate

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$52.40
1 L
$132.00
25 KG
$1,170.00

$52.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$52.40
1 L
$132.00
25 KG
$1,170.00

About This Item

Linear Formula:
(CH3O)3B
CAS Number:
Molecular Weight:
103.91
MDL number:
UNSPSC Code:
12352108
EC Index Number:
204-468-9
NACRES:
NA.22

$52.40


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

147.9-148 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (acidimetric)

form

liquid

autoignition temp.

305 °C

potency

6140 mg/kg LD50, oral (Rat)
1845 mg/kg LD50, skin (Rabbit)

reaction suitability

reagent type: reductant

refractive index

n20/D 1.3568

bp

67-69 °C/1013 hPa

mp

-31 °C

transition temp

flash point -11 °C

density

0.932 g/cm3 at 20 °C

SMILES string

B(OC)(OC)OC

InChI

1S/C3H9BO3/c1-5-4(6-2)7-3/h1-3H3

InChI key

WRECIMRULFAWHA-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Density (d 20 °C/ 4 °C): 0.931 - 0.933
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 1B - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

12.2 °F - (own results)

Flash Point(C)

-11 °C - (own results)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service