Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.21047

Sigma-Aldrich

Methyl propionate

for synthesis

Synonym(s):

Methyl propionate, Propionic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
MDL number:
UNSPSC Code:
12352108
EC Index Number:
209-060-4

vapor pressure

90 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

465 °C

potency

5000 mg/kg LD50, oral (Rat)

expl. lim.

2.5-13 % (v/v)

mp

-87 °C

transition temp

flash point -2 °C

solubility

5 g/L

density

0.91 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C4H8O2/c1-3-4(5)6-2/h3H2,1-2H3

InChI key

RJUFJBKOKNCXHH-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.914 - 0.918
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

28.4 °F - closed cup

Flash Point(C)

-2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service