Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.21026

Sigma-Aldrich

Phosphoryl bromide

for synthesis

Synonym(s):

Phosphoryl bromide, Phosphorus oxide tribromide, Phosphorus oxybromide

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$220.00
100 G
$369.00
1 KG
$1,540.00

$220.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$220.00
100 G
$369.00
1 KG
$1,540.00

About This Item

Empirical Formula (Hill Notation):
Br3OP
CAS Number:
Molecular Weight:
286.69
MDL number:
UNSPSC Code:
12352307
EC Index Number:
232-177-7
NACRES:
NA.22

$220.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

crystals

bp

192 °C/1013 hPa (decomposes)

mp

55 °C

density

2.82 g/cm3

storage temp.

2-30°C

SMILES string

[P](=O)(Br)(Br)Br

InChI

1S/Br3OP/c1-5(2,3)4

InChI key

UXCDUFKZSUBXGM-UHFFFAOYSA-N

Analysis Note

Assay (argentometric): ≥ 98.0 %
Melting range (lower value): ≥ 48 °C
Melting range (upper value): ≤ 56 °C
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service