Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.20933

Sigma-Aldrich

3,5-Dihydroxytoluene

for synthesis

Synonym(s):

3,5-Dihydroxytoluene, 5-Methylresorcinol, Orcinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8O2
CAS Number:
Molecular Weight:
124.14
MDL number:
UNSPSC Code:
12352104
EC Index Number:
207-984-2
NACRES:
NA.22
form:
powder
Assay:
≥99.0% (GC)

Quality Level

Assay

≥99.0% (GC)

form

powder

potency

>2000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

bp

147 °C/7 hPa

mp

109 °C

transition temp

flash point 159 °C

solubility

80 g/L

SMILES string

Oc1cc(cc(c1)C)O

InChI

1S/C7H8O2/c1-5-2-6(8)4-7(9)3-5/h2-4,8-9H,1H3

InChI key

OIPPWFOQEKKFEE-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Water (K. F.): ≤ 4.0 %
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service