Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.20905

Sigma-Aldrich

2-Hydroxy-5-nitrobenzoic acid

for synthesis

Synonym(s):

2-Hydroxy-5-nitrobenzoic acid, 5-Nitrosalicylic acid, 5-Nitro-2-hydroxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(OH)-5-(NO2)C6H3COOH
CAS Number:
Molecular Weight:
183.12
MDL number:
UNSPSC Code:
12352106
EC Index Number:
202-548-8

Quality Level

Assay

≥98.0% (HPLC)

form

powder

mp

229-234 °C

solubility

1.76 g/L

density

1.65 g/cm3 at 25 °C

storage temp.

no temp limit

InChI

1S/C7H5NO5/c9-6-2-1-4(8(12)13)3-5(6)7(10)11/h1-3,9H,(H,10,11)

InChI key

PPDRLQLKHRZIJC-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 229 °C
Melting range (upper value): ≤ 234 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

249.8 °F - open cup

Flash Point(C)

121 °C - open cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service