Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.20801

Sigma-Aldrich

2-Methylhydroquinone

for synthesis

Synonym(s):

2-Methylhydroquinone, 2,5-Dihydroxytoluene, Toluhydroquinone

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$64.60

$64.60


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$64.60

About This Item

Linear Formula:
2-(CH3)C6H3-1,4-(OH)2
CAS Number:
Molecular Weight:
124.14
MDL number:
UNSPSC Code:
12352104
EC Index Number:
202-443-7
NACRES:
NA.22

$64.60


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥97.0% (HPLC)

form

crystals

autoignition temp.

468 °C

potency

200-400 mg/kg LD50, oral (Rat)

mp

124-127 °C

transition temp

flash point 172 °C

solubility

77 g/L

storage temp.

2-30°C

SMILES string

Oc1c(cc(cc1)O)C

InChI

1S/C7H8O2/c1-5-4-6(8)2-3-7(5)9/h2-4,8-9H,1H3

InChI key

CNHDIAIOKMXOLK-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 125 °C
Melting range (upper value): ≤ 128 °C
Identity (IR): passes test

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service