Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.20775

Sigma-Aldrich

Methyl methanesulfonate

for synthesis

Synonym(s):

Methyl methanesulfonate, Methanesulfonic acid methyl ester, MMS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H6O3S
CAS Number:
Molecular Weight:
110.13
MDL number:
UNSPSC Code:
12352103
EC Index Number:
200-625-0

vapor pressure

0.8 hPa ( 59 °C)

Assay

≥99.0% (GC)

form

liquid

potency

225 mg/kg LD50, oral (Rat)

bp

203 °C/1013 hPa

mp

20 °C

transition temp

flash point 104 °C

density

1.30 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C2H6O3S/c1-5-6(2,3)4/h1-2H3

InChI key

MBABOKRGFJTBAE-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.298 - 1.299
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

MILLIPLEX® genotoxicity assays enable high-throughput measurement of DNA damage, reducing time and cost.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service