Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.20753

Sigma-Aldrich

Lithium hydride

for synthesis

Synonym(s):

Lithium hydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HLi
CAS Number:
Molecular Weight:
7.95
MDL number:
UNSPSC Code:
12352306
EC Index Number:
231-484-3

form

powder

potency

78 mg/kg LD50, oral (Rat)

reaction suitability

reagent type: reductant

pH

>7 (20 °C, 21 g/L in H2O)

mp

688 °C

density

0.78 g/cm3 at 20 °C

bulk density

290‑430 kg/m3

storage temp.

2-30°C

InChI

1S/Li.H/q+1;-1

InChI key

SRTHRWZAMDZJOS-UHFFFAOYSA-N

Analysis Note

Assay (by gasvolumetry): ≥ 96.0 %

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B - Water-react. 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials, which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service