Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.20699

Sigma-Aldrich

1-Indanone

for synthesis

Synonym(s):

1-Indanone, α-Hydrindone, 1-Oxoindane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8O
CAS Number:
Molecular Weight:
132.16
MDL number:
UNSPSC Code:
12352115
EC Index Number:
201-470-1
Pricing and availability is not currently available.

Assay

≥98.0% (GC)

autoignition temp.

525 °C

bp

243-245 °C

mp

37-40 °C

transition temp

flash point 113 °C

storage temp.

2-30°C

InChI

1S/C9H8O/c10-9-6-5-7-3-1-2-4-8(7)9/h1-4H,5-6H2

InChI key

QNXSIUBBGPHDDE-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 39 °C
Melting range (upper value): ≤ 42 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

233.0 °F - closed cup

Flash Point(C)

111.67 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service