Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.20664

Sigma-Aldrich

3-Hydroxy-2-butanone

for synthesis

Synonym(s):

3-Hydroxy-2-butanone, Acetyl methyl carbinol, Acetoin

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$104.00
250 G
$232.00

$104.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$104.00
250 G
$232.00

About This Item

Linear Formula:
(CH3CH(OH)COCH3)2
CAS Number:
Molecular Weight:
176.21
MDL number:
UNSPSC Code:
12352115
NACRES:
NA.22

$104.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥97.0% (GC)

form

liquid

potency

>5000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

bp

147-148 °C/1013 hPa

mp

20 °C (Above 20°C conversion of dimer to monomer 15°C)

transition temp

flash point 45 °C

density

1.012 g/cm3 at 20 °C

SMILES string

OC(C)C(=O)C

InChI

1S/C4H8O2/c1-3(5)4(2)6/h3,5H,1-2H3

InChI key

ROWKJAVDOGWPAT-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Identity (IR): passes test

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

113.0 °F

Flash Point(C)

45 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service