Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.20560

Sigma-Aldrich

Isopropyl acetate

for synthesis

Synonym(s):

Isopropyl acetate

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$35.40
1 L
$51.40
2.5 L
$95.10
175 KG
$2,260.00
170 KG
$3,360.00

$35.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$35.40
1 L
$51.40
2.5 L
$95.10
175 KG
$2,260.00
170 KG
$3,360.00

About This Item

Linear Formula:
CH3COOCH(CH3)2
CAS Number:
Molecular Weight:
102.13
MDL number:
UNSPSC Code:
12352108
EC Index Number:
203-561-1
NACRES:
NA.22

$35.40


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

61 hPa ( 20 °C)

Quality Level

Assay

≥99% (GC)

form

liquid

autoignition temp.

460 °C

potency

6750 mg/kg LD50, oral (Rat)
>20000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.8-8 % (v/v)

bp

88-90 °C/1013 hPa

mp

-73 °C

transition temp

flash point 4 °C

density

0.87 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C(C)C)C(=O)C

InChI

1S/C5H10O2/c1-4(2)7-5(3)6/h4H,1-3H3

InChI key

JMMWKPVZQRWMSS-UHFFFAOYSA-N

Application

Isopropyl acetate can be used as an:
  • Acyl donor in stereoselective biotransformations of chiral amines[1].
  • Alkylating agent for the isopropylation of phenols due to its easy chemisorption on the Bronsted acid sites to form alkyl cations and the release of acetic acid in the reaction that can stabilize the charged structure of phenol[2].

Analysis Note

Assay (GC, area%): ≥ 99 %
Density (d 20 °C/ 4 °C): 0.871 - 0.873
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

41.0 °F

Flash Point(C)

5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isopropyl 2-ethoxyacetate-an efficient acylating agent for lipase-catalyzed kinetic resolution of amines in batch and continuous-flow modes
Olah M, et al.
Tetrahedron, 72, 7249-7255 (2016)
Catalytic performance of Al-MCM-48 molecular sieves for isopropylation of phenol with isopropyl acetate
Venkatachalam K, et al.
Chinese Journal of Catalysis, 33, 478-486 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service